missing translation for 'onlineSavingsMsg'
Learn More

Prolactin Ab-2 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Prolactin Ab-2, Mouse Monoclonal Antibody.

Brand:  Epredia MS-1362-P1

 View more versions of this product

Product Code. 12629656

  • ONLINE EXCLUSIVE PRICE
    435.00€
    363.83€ / 500µL
    Save 71.17€
    16% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Prolactin is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. Prolactin producing cells make up approximately 20% of the pituitary.

Host Species: Mouse

Clone: PRL02

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human prolactin

Molecular Weight: 23.5kDa

Positive Control: Pituitary gland

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • ELISA

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Prolactin Ab-2
Monoclonal
200μg/mL
Mouse
500 μL
Steroids Hormones and Growth Factors
Human
IgG2a κ
Immunohistochemistry (Paraffin)
PRL02
Unconjugated
Human prolactin
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.