Learn More
Labconco™ Moisture Monitor/Hygrometer
Product Code. 12697755 Shop All Labconco Corporation Products
Change view
Click to view available options
Electrical Requirements:
230 V 50/60 Hz
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Electrical Requirements unitSize
12697755 230 V 50/60 Hz Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 12697755 Supplier Labconco™ Supplier No. 5244901
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
For placement inside the glove box main chamber to continuously monitor moisture levels
  • Measures absolute moisture content in gas
  • Detects levels from 0 to 10,000ppm (with three ranges of 0-100ppm, 100-1000ppm, and 1000-10,000ppm) with ±1 percent accuracy
  • Consists of chemically-inert ceramic technology moisture sensor, sampling block, digital display monitor, sampling pump, switching power supply and on/off switch
  • For use with all glove boxes
Specifications
Type Moisture Monitor/Hygrometer
Resolution 5 ppm
Accuracy ±1%
Length (English) 7.7 in.
Height (Metric) 18.28 cm
Electrical Requirements 230 V 50/60 Hz
For Use With (Application) For placement inside the glove box main chamber to continuously monitor moisture levels