Learn More
Labconco™ Protector Glove Box Accessory Kits
Product Code. 16649183 Shop All Labconco Corporation Products
Click to view available options
Product Type:
Pressure Relief Kit
Unit Size:
Each
Product Code. 16649183
Brand: Labconco™ 5441403
Kits provided added functionality options for Protector glove boxes
Kit Components
  • 5 in. diameter round HEPA filter (99.9% efficient on 0.3 micron particulates)
  • Plastic hose barbs (2)
  • 10 ft. of 3/4 in. ID neoprent tubing
Specifications
For Use With (Equipment) Protector Glove Boxes
Includes HEPA filter, hose barb connectors and 10 ft. of 0.75 in. ID neoprene tubing for attachment to the pressure relief bubbler exhaust
Product Type Pressure Relief Kit