Learn More
Labconco™ Secondary Trap Canister and Inserts for CentriVap™ System
Product Code. 10405395 Shop All Labconco Corporation Products
Change view
Click to view available options
Type:
Acid Trap Insert
Moisture Trap Insert
Radioisotope Trap Insert
Secondary Trap Canister
Solvent Trap Insert
Weight (Metric):
1.8 kg
2.3 kg
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Weight (Metric) unitSize
10405395 Secondary Trap Canister 2.3 kg Each
15173095 Radioisotope Trap Insert 1.8 kg Each
11761479 Solvent Trap Insert 1.8 kg Each
10064023 Acid Trap Insert 1.8 kg Each
13265130 Moisture Trap Insert 1.8 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10405395 Supplier Labconco™ Supplier No. 7460900
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket
Help to provide secondary protection to vacuum pumps in evaporation systems
  • Clear polypropylene canister with mounting bracket, 17 in. (43cm) PVC tubing, and two clamps
  • Canister accommodates trap inserts listed in the table
Specifications
Color Clear
Material Polypropylene
For Use With (Equipment) CentriVap™ system
Includes Mounting bracket
Type Secondary Trap Canister
Weight (English) 5 lb.
Weight (Metric) 2.3 kg