Learn More
Brady™ Label Printer Labels
Self-laminating polyester for printer.
Brand: Brady™ 710776
Additional Details : Weight : 0.12972kg
Description
- Made from self-laminating polyester
- Abrasion- and smudge-resistant
- Permanent Acrylic adhesive
- White/Transparent
Specifications
BMP71 Label Printer Labels | |
White/Transparent | |
Self-laminating | |
Printer Label | |
1 | |
Rectangle | |
0.048 mm | |
15.24 mm |
Permanent Acrylic | |
Matte | |
Polyester | |
Laboratory identification | |
500 Labels/Box | |
15.24 x 41.28 mm (W x H) | |
41.28 x 15.24 mm | |
41.28 mm |