Learn More
SHP Steriltechnik AG Laboklav 25 Benchtop Autoclave
Product Code. 31908743
Change view
Click to view available options
Capacity (Metric):
25 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
31908743 25 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Regulatory: This product is under eco-contribution tax.
Product Code. 31908743 Supplier SHP Steriltechnik AG Supplier No. 21.0025.001.000
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Regulatory: This product is under eco-contribution tax.
Benchtop autoclave designed for laboratory sterilisation of instruments, liquids, and waste.
  • Benchtop device with 25 litre chamber size
  • Usable space (Ø x L) 265 x 465 mm, fits up to 5 x 1 L laboratory bottles or 5 kg of instruments/waste
  • Operating overpressure 2.8/-1 bar
  • Sterilisation temperature 98 °C to 135 °C
  • 2.0 kW heating power, 230 V~, 50 Hz, 13 A
  • Integrated steam generator and automatic feed water pump
  • Safety valve, TÜV-approved component
  • Motorised closing mechanism with lip seal
  • Chamber lid made of safety glass with illuminated see-through area
  • Door cover made of safety glass with coloured LED lighting
  • Thermo locking device with flexible media sensor as standard
  • Fully automatic control with up to 10 programmes, customizable and code-secured
  • Integrated exhaust vapour condensation
  • Integrated feed water supply tank and condensate tank
  • RS-485 interface
  • 4 MB internal memory chip, stores up to 150 cycle data
Specifications
Capacity (Metric) 25 L
Depth (Metric) Chamber 450 mm
Product Type Horizontal, Front-loading Benchtop Autoclave