Learn More
Smeg™ Laboratory Glassware Washer
Product Code. 11932699
Change view
Click to view available options
Includes:
20 default programs + 10 free
20 default programs + 10 free, active thermodynamic drying
Electrical Requirements:
3/N/PE 400V - 50Hz - 7 kW max (switchable single-phase 230V)
Temperature Setting:
Washing up to 95°C
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Electrical Requirements Temperature Setting unitSize
11932699 20 default programs + 10 free 3/N/PE 400V - 50Hz - 7 kW max (switchable single-phase 230V) Washing up to 95°C Each
11942699 20 default programs + 10 free, active thermodynamic drying 3/N/PE 400V - 50Hz - 7 kW max (switchable single-phase 230V) Washing up to 95°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11932699 Supplier Smeg™ Supplier No. GW3060S
  • ONLINE EXCLUSIVE PRICE
    10210.81€
    8500.00€ / Pack of 1
    Save 1710.81€
    16% Off
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Clean a wide range of glassware with this washer. Smeg™ Laboratory Glassware Washers are manufactured with high quality materials and feature faultless, intuitive performance. Specific accessories ensure high quality washing of even the most difficult and dirtier instruments.
  • Microprocessor electronic control
  • Active thermodynamic drying
  • Alkaline powder detergent dispenser
  • Peristaltic pump for acid liquid neutralizer
  • Washing chamber and internal door in AISI 304 stainless steel
  • External panels made in AISI 304 stainless steel
  • 2 independent washing levels through telescopic guides
  • Incorporated water softener with salt sensor
  • Cold water and demineralized water connections
  • Electronic door lock system for user safety
  • Without trolleys
Specifications
Material Stainless Steel
Product Type Glassware Washer
Includes 20 default programs + 10 free
Temperature Setting Washing up to 95°C
Dimensions (D x W x H) Interior 490 x 525 x 570mm
Electrical Requirements 3/N/PE 400V - 50Hz - 7 kW max (switchable single-phase 230V)