missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LACTB2 Monoclonal Antibody (OTI1A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
242.00€ - 540.87€
Specifications
Antigen | LACTB2 |
---|---|
Clone | OTI1A8 |
Concentration | 1 mg/mL |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17537271
|
Origene Technologies
TA502585S |
30 μL |
242.00€
30µL |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17517271
|
Origene Technologies
TA502585 |
100 μL |
540.87€
100µL |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
LACTB2 Monoclonal antibody specifically detects LACTB2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotEspecificaciones
LACTB2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Q53H82 | |
IgG2b | |
-20° C, Avoid Freeze/Thaw Cycles |
OTI1A8 | |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
51110 | |
Primary |