missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LACTB2 Monoclonal Antibody (OTI4F7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502589
Additional Details : Weight : 0.05000kg
Description
Endoribonuclease; cleaves preferentially 3′ to purine-pyrimidine dinucleotide motifs in single-stranded RNA. The cleavage product contains a free 3′ -OH group. Has no activity with double-stranded RNA or DNA. Required for normal mitochondrial function and cell viability.Specifications
LACTB2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q53H82 | |
LACTB2 | |
Full length human recombit protein of human LACTB2 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI4F7 | |
Unconjugated | |
LACTB2 | |
CGI-83 | |
Mouse | |
Affinity Chromatography | |
RUO | |
51110 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |