missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LACTB2 Mouse anti-Human, Clone: OTI1A8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF502585
Additional Details : Weight : 0.05000kg
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Endoribonuclease; cleaves preferentially 3′ to purine-pyrimidine dinucleotide motifs in single-stranded RNA. The cleavage product contains a free 3′ -OH group. Has no activity with double-stranded RNA or DNA. Required for normal mitochondrial function and cell viability.Specifications
LACTB2 | |
Monoclonal | |
Unconjugated | |
LACTB2 | |
CGI-83 | |
Mouse | |
Affinity Chromatography | |
RUO | |
51110 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1A8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q53H82 | |
LACTB2 | |
Full length human recombit protein of human LACTB2 produced in E.coli | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |