missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid Dodecyl Ester, TRC

Product Code. 30138495
Change view
Click to view available options
Quantity:
100 mg
500 mg
5 g
Unit Size:
100mg
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138495 5 g 5g
30135250 100 mg 100mg
30005245 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138495 Supplier Toronto Research Chemicals Supplier No. TRCL1135105G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6283-92-7
Molecular Formula C15 H30 O3
Molecular Weight (g/mol) 258.4
Synonym Lactic acid, dodecyl ester (7CI), Ceraphyl 31, Chrystap HYL 98, Crodamol LL, Cyclochem LVL, Dodecyl lactate, Lauryl lactate, NSC 7752, Pelemol LL, Purasolv LL, Schercemol LL, Lactic Acid-Dodecyl Ester
IUPAC Name dodecyl 2-hydroxypropanoate
SMILES CCCCCCCCCCCCOC(=O)C(C)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C15H30O3/c1-3-4-5-6-7-8-9-10-11-12-13-18-15(17)14(2)16/h14,16H,3-13H2,1-2H3
Formula Weight 258.2195
Chemical Name or Material Lactic Acid Dodecyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.