Learn More
Lactitol monohydrate, 98%
Product Code. 11345349
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11345349 1 g 1g
11359699 5 g 5g
2 options
Product Code. 11345349
Brand: Thermo Scientific Alfa Aesar B21016.03
Request Bulk or Custom Format
CAS: 81025-04-9 | C12H26O12 | 362.328 g/mol
Lactitol monohydrate is used in low fat foods, anthelmintic, cosmetic and pharmaceutical excipients. It is used in sugar-free candies, cookies, chocolate and ice cream. It is a laxative, which used to treat constipation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Lactitol monohydrate is used in low fat foods, anthelmintic, cosmetic and pharmaceutical excipients. It is used in sugar-free candies, cookies, chocolate and ice cream. It is a laxative, which used to treat constipation.
Solubility
Soluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 81025-04-9
Molecular Formula C12H26O12
Molecular Weight (g/mol) 362.328
MDL Number MFCD00150767
InChI Key LXMBXZRLTPSWCR-XBLONOLSSA-N
Synonym lactitol monohydrate, d-lactitol monohydrate, lactitol hydrate, unii-uh2k6w1y64, 4-o-beta-d-galactopyranosyl-d-glucitol monohydrate, d-glucitol, 4-o-beta-d-galactopyranosyl-, monohydrate, 4-o-beta-d-galactopyranosyl-d-glucitol, portolac tn, 2s,3r,4r,5r-4-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxyhexane-1,2,3,5,6-pentol;hydrate
PubChem CID 3067270
IUPAC Name (2S,3R,4R,5R)-4-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,5,6-pentol;hydrate
SMILES C(C1C(C(C(C(O1)OC(C(CO)O)C(C(CO)O)O)O)O)O)O.O
Specifications
Melting Point 95°C to 98°C
Quantity 1 g
Merck Index 14,5340
Solubility Information Soluble in water.
Formula Weight 362.33 (344.31 Anhydrous)
Percent Purity 98%
Chemical Name or Material Lactitol monohydrate