missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactitol Monohydrate, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | D-Lactitol monohydrate |
| CAS | 81025-04-9 |
| Molecular Formula | C12 H24 O11 . H2 O |
| Quantity | 1 g |
| Synonym | Lactitol monohydrate, 4-O-(β-d-Galactopyranosyl)-d-glucitol monohydrate |
| InChI Formula | InChI=1S/C12H24O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h4-21H,1-3H2;1H2/t4-,5+,6+,7+,8-,9-,10+,11+,12-;/m0./s1 |
| SMILES | O.OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)CO |
| IUPAC Name | (2S,3R,4R,5R)-4-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,5,6-pentol;hydrate |
| Molecular Weight (g/mol) | 362.33 |
| Formula Weight | 362.1424 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction