missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactoferrin Antibody (LF5-1D2), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-67260H
This item is not returnable.
View return policy
Description
Lactoferrin Monoclonal specifically detects Lactoferrin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Lactoferrin | |
| Monoclonal | |
| 1mg/mL | |
| Western Blot, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin | |
| Mouse | |
| Protein A purified | |
| Amino Acids Drugs and other small molecules | |
| 4057 | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| LF5-1D2 | |
| HRP | |
| PBS with No Preservative | |
| LTF | |
| Human lactoferrin | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction