missing translation for 'onlineSavingsMsg'
Learn More

Lactose Monohydrate, Extra Pure, SLR, Fine Powder, Fisher Chemical™

Product Code. 10497492 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10497492 500 g Plastic powder jar 500g
10427070 1 kg Plastic powder jar 1kg
10151430 5 kg Plastic keg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10497492 Supplier Fisher Chemical Supplier No. L/0200/53

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 63-42-3
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number 71587
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
Synonym d-lactose monohydrate, d-+-lactosemonohydrate
PubChem CID 133126686
IUPAC Name (3S,6R)-2-(hydroxymethyl)-6-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

Melting Point 219°C
Color White
Packaging Plastic powder jar
Quantity 500 g
Formula Weight 360.3g/mol
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.