missing translation for 'onlineSavingsMsg'
Learn More

Lactose Monohydrate, Mikromol™

Product Code. 17949244
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17949244 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17949244 Supplier Mikromol Supplier No. MM1485.00
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Lactose Monohydrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha-Lactose Monohydrate
CAS 5989-81-1
Analyte or Component Names alpha-Lactose Monohydrate
Shipping Condition Room Temperature
Molecular Formula C12 H22 O11 . H2 O
Certifications/Compliance ISO 17034
Synonym Aeroflo 20, Aeroflo 65, Aeroflo 95, Flowlac 200, GranuLac 200, Granulac 230, Inhalac 120, Inhalac 230, Lactochem, Lactose 100M, Lactose D 80, Lactosum D 80, Pharmatose 150M, Pharmatose 325, Pharmatose 50M, Pharmatose 90M, Pharmatose DCL 11, Zeparox, 4-O-beta-D-Galactopyranosyl-alpha-D-glucopyranose monohydrate, alpha-D-Lactose monohydrate, alpha-Lactose monohydrate
InChI Formula InChI=1S/C12H22O11.H2O/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17;/h3-20H,1-2H2;1H2/t3-,4-,5+,6+,7-,8-,9-,10-,11+,12+;/m1./s1
SMILES O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
Molecular Weight (g/mol) 360.31
Quantity 250 mg
Formula Weight 360.1268 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.