Learn More
Serva Electrophoresis™ Laemmli Sample Buffer, 2X
Product Code. 11702177
Encompass_Preferred
Change view
Click to view available options
Quantity:
20mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11702177 20mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11702177 Supplier Serva Electrophoresis™ Supplier No. 4252601
 Restricted Item
Estimated Shipment: 11-08-2026
Specifications
Concentration 2X
For Use With (Application) SDS-PAGE
Material 126mM Tris/Cl (pH 6.8), 20% glycerol, 4% SDS and 0.02% bromophenol blue
Quantity 20mL