missing translation for 'onlineSavingsMsg'
Learn More

LAG-3 Antibody (L4-PL33) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-23442-100ug

 View more versions of this product

Product Code. 18612664

  • 658.00€ / 100µg
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LAG-3 Monoclonal antibody specifically detects LAG-3 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Neutralization
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LAG-3
Monoclonal
1 mg/ml
Western Blot 5 ug/mL, Flow Cytometry, ELISA, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 10 ug/mL, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:150, Block/Neutralize
17b4, 17b4 lag3, 17b4 neutralizing, anti human lag3, Anti-LAG-3, CD223, CD223 Antigen, Human LAG3, Human LAG-3, LAG3, LAG-3, LAG-3 17b4, LAG-3 blocking, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3
This LAG-3 Antibody (L4-PL33) was prepared from synthetic peptide corresponding to 30 aa (GPPAAAPGHPLAPGPHPAAPSSWGPRPRRY) from the first N-terminal D1 domain of human LAG-3 (lymphocyte activation gene-3).
100 μg
Adaptive Immunity, Cancer, Immunology, Innate Immunity
3902
Store at 4°C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1 κ
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
L4-PL33
Unconjugated
PBS, 10% glycerol
Mouse
Protein A or G purified
RUO
Primary
Human, Primate
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.