missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lambda Light Chain Antibody (SR1399), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00€ - 449.00€
Specifications
| Antigen | Lambda Light Chain |
|---|---|
| Clone | SR1399 |
| Dilution | Western Blot 1:1000-1:5000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18672755
|
Novus Biologicals
NBP3-21819-100ul |
100 μg |
449.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18614044
|
Novus Biologicals
NBP3-21819-50ul |
50 μg |
338.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lambda Light Chain Monoclonal antibody specifically detects Lambda Light Chain in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
| Lambda Light Chain | |
| Western Blot 1:1000-1:5000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| IGLC, immunoglobulin lambda constant group | |
| A synthesized peptide derived from human Lambda Light Chain (Uniprot #: P01701, P01842) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1399 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Immunology | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 3537 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title