missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMC2 Antibody (CL2980) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42388UV
This item is not returnable.
View return policy
Description
LAMC2 Monoclonal antibody specifically detects LAMC2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| LAMC2 | |
| Monoclonal | |
| DyLight 350 | |
| BM600, BM600-100kDa, Cell-scattering factor 140 kDa subunit, CSF, CSF 140 kDa subunit, EBR2, EBR2A, Epiligrin subunit gamma, kalinin (105kD), Kalinin subunit gamma, Kalinin/nicein/epiligrin 100 kDa subunit, kalinin-105kDa, ladsin (140kDa), Ladsin 140 kDa subunit, LAMB2Tlaminin, gamma 2 (nicein (100kD), kalinin (105kD), BM600 (100kD), Herlitzjunctional epidermolysis bullosa)), Laminin B2t chain, laminin subunit gamma-2, laminin, gamma 2, Laminin-5 subunit gamma, LAMNB2B2T, Large adhesive scatter factor 140 kDa subunit, MGC138491, MGC141938, nicein (100kDa), Nicein subunit gamma, nicein-100kDa | |
| This antibody was developed using a recombinant protein derived from Q13753, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| 3918 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL2980 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction