missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lamin B Receptor Antibody (BB2SS3F3), Novus Biologicals™
Mouse Monoclonal Antibody
214.00€ - 471.00€
Specifications
| Antigen | Lamin B Receptor |
|---|---|
| Clone | BB2SS3F3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1-2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18615338
|
Novus Biologicals
NBP2-59947 |
0.1 mg |
471.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18686667
|
Novus Biologicals
NBP2-59947-0.025mg |
0.025 mg |
214.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lamin B Receptor Monoclonal antibody specifically detects Lamin B Receptor in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
| Lamin B Receptor | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Cellular Signaling | |
| PBS | |
| 3930 | |
| IgG1 κ | |
| Protein A purified |
| BB2SS3F3 | |
| Western Blot 1:100 - 1-2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| DHCR14B, FLJ43126, Integral nuclear envelope inner membrane protein, lamin B receptor, lamin-B receptor, LMN2R, MGC9041, PHA | |
| GST fused to N-terminal fragment of human Lamin B Receptor (amino acids residues 1-211) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title