Learn More
Brady™ Laminated Polyester ADR 1.5 1 Explosive Substances Transport Signs
Product Code. 11760204 Shop All Brady Products
Change view
Click to view available options
Height (Metric):
100 mm
200 mm
297 mm
Width (Metric):
100.00 mm
200.00 mm
297.00 mm
Quantity:
1 Ea.
250/Pk.
Unit Size:
1 roll
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Width (Metric) Quantity Unit Size
11760204 297 mm 297.00 mm 1 Ea. Each
11740204 100 mm 100.00 mm 1 Ea. Each
11790234 100 mm 100.00 mm 250/Pk. 1 roll
11750204 200 mm 200.00 mm 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11760204 Supplier Brady™ Supplier No. 223557
  • ONLINE EXCLUSIVE PRICE
    10.85€
    9.36€ / Pack of 1
    Save 1.49€
    13% Off
Estimated Shipment: 12-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Alternate Part Number ADR 1.5-297*297-B7541
Finish Gloss
Material Laminated Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Height (Metric) 297 mm
Thickness 0.099 mm
Width (Metric) 297.00 mm
Temperature Range (Metric) -40 °C-110 °C
Shape Diamond
Displayed Text 1.5 1
Color Black on Orange