Learn More
Brady™ Laminated Polyester ADR 5.1 Oxidizing Substance Transport Signs
Product Code. 11760224 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
100 x 100 mm
Unit Size:
1 roll
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
11760224 100 x 100 mm 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11760224 Supplier Brady™ Supplier No. 223580
  • ONLINE EXCLUSIVE PRICE
    398.00€
    359.51€ / 1 rolls
    Save 38.49€
    9% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Alternate Part Number ADR 5.1RL-100*100-B7541
Finish Gloss
Material Laminated Polyester
Quantity 250/Pk.
Adhesive Type Permanent Acrylic
Height (Metric) 100 mm
Thickness 0.099 mm
Width (Metric) 100.00 mm
Temperature Range (Metric) -40 °C-110 °C
Shape Diamond
Dimensions (L x W) 100 x 100 mm
Displayed Text 5.1
Color Black on Yellow