missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin alpha 3 Antibody (CL3112) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42390DL594
This item is not returnable.
View return policy
Description
Laminin alpha 3 Monoclonal antibody specifically detects Laminin alpha 3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Laminin alpha 3 | |
| Monoclonal | |
| DyLight 594 | |
| BM600, BM600 150kD subunit, BM600-150kDa, E170, epiligrin, Epiligrin 170 kDa subunit, epiligrin alpha 3 subunit, Epiligrin subunit alpha, kalinin 165kD subunit, Kalinin subunit alpha, kalinin-165kDa, lama3a, laminin subunit alpha-3, laminin, alpha 3, laminin, alpha 3 (nicein (150kD), kalinin (165kD), BM600 (150kD), epilegrin), laminin-5 alpha 3 chain, Laminin-5 subunit alpha, Laminin-6 subunit alpha, Laminin-7 subunit alpha, LAMNA, LOCS, nicein 150kD subunit, Nicein subunit alpha, nicein-150kDa | |
| This antibody was developed using a recombinant protein derived from Q16787, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Signal Transduction | |
| 3909 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL3112 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction