missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin alpha 4 Antibody (CL3185) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42393MFV500
This item is not returnable.
View return policy
Description
Laminin alpha 4 Monoclonal antibody specifically detects Laminin alpha 4 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Laminin alpha 4 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| DKFZp686D23145, LAMA3LAMA4*-1, laminin alpha 4 chain, laminin subunit alpha-4, laminin, alpha 4, Laminin-14 subunit alpha, Laminin-8 subunit alpha, Laminin-9 subunit alpha | |
| This antibody was developed using a recombinant protein derived from Q16363, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL3185 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3910 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction