Learn More
Invitrogen™ Laminin beta-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582138
Description
Immunogen sequence: NQVVSLSPGS RYVVLPRPVC FEKGTNYTVR LELPQYTSSD SDVESPYTLI DSLVLMPYCK SLDIFTVGGS GDGVVTNSAW ETFQRYRCLE NSRSVVKTPM TDVCRNIIFS ISALLHQTGL ACECDPQGSL SSVCDPNGG Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 96%.
Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.
Specifications
| Laminin beta-1 | |
| Polyclonal | |
| Unconjugated | |
| LAMB1 | |
| C77966; C80098; C81607; CLM; D130003D08Rik; I79_017505; LAMB1; Lamb-1; LAMB1; basement membrane protein; Lamb1-1; laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin beta 1; laminin beta-1 chain; laminin subunit beta 1; laminin subunit beta-1; laminin, beta 1; laminin-1 subunit beta; Laminin-10 subunit beta; laminin-12 subunit beta; Laminin-2 subunit beta; laminin-6 subunit beta; Laminin-8 subunit beta; LIS5; LOW QUALITY PROTEIN: laminin subunit beta-1; MGC142015 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3912 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P07942 | |
| LAMB1 | |
| Recombinant protein corresponding to Human LAMB1. Recombinant protein control fragment (Product #RP-102389). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.