missing translation for 'onlineSavingsMsg'
Learn More

Lamotrigine, LoGiCal

Product Code. 30462894
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462894 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462894 Supplier LoGiCal Supplier No. LGCFOR0922.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lamotrigine
CAS 84057-84-1
Analyte or Component Names Lamotrigine
Shipping Condition Room Temperature
Molecular Formula C9 H7 Cl2 N5
Certifications/Compliance ISO 9001
Synonym Lamotrigine, 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, 3,5-Diamino-6-(2,3-dichlorophenyl)-1,2,4-triazine, BW 430C, LTG, Lamictal, Lamotrigin, 430C78, GI 267119X, 3,4-stage 2 isomer, Lamotrigine 430C78, 13W80 (Ph Eur Lamotrigine Impurity B), Lamotrigine stage 2 14W80 (Ph Eur Lamotrigine Impurity C)
InChI Formula InChI=1S/C9H7Cl2N5/c10-5-3-1-2-4(6(5)11)7-8(12)14-9(13)16-15-7/h1-3H,(H4,12,13,14,16)
Solution Type Neat
SMILES Nc1nnc(c(N)n1)c2cccc(Cl)c2Cl
IUPAC Name 6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine
Industry Type Clinical Testing
Molecular Weight (g/mol) 256.09
Quantity 10 mg
Formula Weight 255.0079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.