Learn More
Logistik™ Lancer-Style Chemical Splash Laboratory Coats
Product Code. 11999155
Change view
Click to view available options
Color: white; Size: L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11999155 Color: white; Size: L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11999155 Supplier Logistik™ Supplier No. CS849WHL
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Provide the answer to full frontal protection with Logistik™ Lancer-Style Chemical Splash Laboratory Coats The wrap-over style Lancer front coat helps repel liquid chemicals and provides the wearer with an excellent level of protection since the use of chemicals in a laboratory can prove a constant hazard to standard style open neck coats.
  • Full frontal protection
  • Chem-Tech material
  • Fluorochemical liquid repellent
Specifications
Product Type General Purpose Lab Coat
Size Large
Gender Unisex
Material Polyester, Carbon
Color White
Cuff Style Stud-fastened
Disposable Reusable
Sterility Non-sterile
Style Lancer
No. of Pockets 2
Collar Type Open
Finish Fluorochemical Liquid Repellent
Size (Metric) Chest 106.68 to 111.76 cm
Certifications/Compliance CE EN 13034
Length (Metric) 108 cm
Construction 65% Polyester, 35% Cotton