missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum(III) chloride heptahydrate, Puriss. p.a., ≥99%, Solstice
Shop All Honeywell Fluka ProductsDescription
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 100 g |
| Molecular Formula | Cl3H14LaO7 |
| Linear Formula | LaCl3 · 7H2O |
| MDL Number | MFCD00149756 |
| UN Number | NONH for all modes of transport |
| InChI Key | FDFPDGIMPRFRJP-UHFFFAOYSA-K |
| SMILES | O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3] |
| IUPAC Name | lanthanum(3+) heptahydrate trichloride |
| Molecular Weight (g/mol) | 371.36 |
| Formula Weight | 371.37g/mol |
| Show More |
Safety and Handling
Product Identifier
- Lanthanum(III) chloride heptahydrate
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin sensitiser Category 1
- H290-May be corrosive to metals.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- P234-Keep only in original container.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: Lanthanum chloride heptahydrate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?