missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-AKT [pSer473] Antibody
AKT is phosphorylated at Ser473 by the rapamycin-insensitive mTor complex (mTORC2).
4360.00€ - 24150.00€
Specifications
| Conjugate | Tb (Terbium)/Lanthanide Chelates |
|---|---|
| Excitation/Emission | 340/490, 520 |
| Product Line | LanthaScreen |
| Reactivity | Human |
| Readout | End Point |
Description
- AKT is phosphorylated at Ser473 by the rapamycin-insensitive mTor complex (mTORC2)
- For use in combination with the LanthaScreen™ AKT HEK293E cell line (K1516) to evaluate inhibitors in medium to high throughput screening applications
Specifications
| Tb (Terbium)/Lanthanide Chelates | |
| LanthaScreen | |
| End Point | |
| Ser/Thr Kinases |
| 340/490, 520 | |
| Human | |
| Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title