Learn More
Thermo Scientific™ LanthaScreen™ Tb-p-c-jun (pSer73) Antibody
For use in combination with the GFP-c-Jun physiological protein substrate (PV4446) to evaluate inhibitors in medium to high throughput screening applications.
Brand:  Thermo Scientific™ PV4450
 View more versions of this product
Product Code. 10155164
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • A part of the activating protein-1 (AP-1) transcription factor, c-Jun is phosphorylated at Ser73 by members of the JNK family
  • For use in combination with the GFP-c-Jun physiological protein substrate (PV4446) to evaluate inhibitors in medium to high throughput screening applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tb (Terbium)/Lanthanide Chelates
Terbium-labeled p-c-Jun (pSer73) antibody in HEPES buffered saline. Store at -20°C.
Human
End Point
Ser/Thr Kinases
1 mg
340/490, 520
Dry Ice
Hu
LanthaScreen
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.