Learn More
Thermo Scientific™ LanthaScreen™ Tb-pATF2 (pSer71) Antibody Kit
Product Code. 11525046
Change view
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
1mg
25µg
Product Code. Quantity unitSize
11525046 25 μg 25µg
10175772 1 mg 1mg
2 options
Product Code. 11525046
Brand: Thermo Scientific™ PV4451
LanthaScreen™ Tb-pATF2 (pSer71) Antibody Kit
A member of the leucine zipper family of DNA binding proteins, ATF2 (activating transcription factor 2) is phosphorylated at Ser71 by members of the JNK and p38 families of kinases.
For use with the GFP-ATF2 physiological protein substrate (PV4445) to evaluate inhibitors in medium to high throughput screening applications.
TRUSTED_SUSTAINABILITY
Specifications
Antigen Serine/threonine kinase
Applications Kinase Assay, Enzyme and Protein Activity Assays, Kinase Assay
Conjugate Tb (Terbium)/GFP (EmGFP)
Description LanthaScreen™ Tb-pATF2 Antibody Kit, 25μg, 150kDa Molecular Weight, TR-FRET Technique, Ser/Thr kinases Antigen, Human Species Reactivity, GFP (EmGFP), Tb (Terbium) Conjugate, Used For Biochemical Kinase Assays, Drug Discovery and Development
Formulation 384-well plate
Quantity 25 μg
Regulatory Status RUO
Research Discipline Protein Biology
Primary or Secondary Primary
Packaging 1 Kit(s)
Content And Storage Terbium-labeled anti-ATF2 [pSer71] antibody in HEPES buffered saline. Store at -20°C.
Antibody Molecular Weight 150000 Da
Assay Entry Biochemical terbium phospho-specific activity
Format 384-well Plate
Excitation/Emission 340⁄490, 520
Reactivity Human
Shipping Condition Dry Ice
Readout End Point
Reactivity abrv Hu
Target Entry MAPK13, MAPK8, MAPK9, MAPK14, MAPK3, MAPK1, MAPK11, MAPK12, MAPK10
Target Enzyme Serine⁄Threonine Kinases
Technique TR-FRET
Validated Kinases MAPK1, MAPK3, MAPK8, MAPK9, MAPK10, MAPK11, MAPK12, MAPK13, MAPK14
Product Line LanthaScreen