missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ TR-FRET RXR beta Coactivator Assay Kit, goat
Description
- This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15120:
- The LanthaScreen™ TR-FRET Retinoid X Receptor (RXR) beta Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential RXR beta ligands as agonists of coactivator recruitment. The homogeneous mix-and-read assay format results in a ligand EC50 composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and recruit coactivator peptide (Figure 1).
- The LanthaScreen™ TR-FRET RXR beta Coactivator Assay Kit includes a terbium (Tb)-labeled anti-GST antibody, a fluorescein-labeled coactivator peptide, and a human RXR beta ligand-binding domain (RXR beta-LBD) that is tagged with glutathione-S-transferase (GST). To assay, the RXR beta-LBD is added to ligand test compounds, followed by addition of a mixture of the fluorescein-coactivator peptide and Tb-anti-GST antibody. After room temperature incubation, the TR-FRET 520:495 emission ratio is calculated and used to determine the EC50 from a dose response curve of the compound
- The LanthaScreen™ TR-FRET RXR beta Coactivator Assay Kit contains RXR beta-LBD (GST) protein, fluorescein-labeled D22 coactivator peptide, Tb-anti-GST antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C).
Specifications
Specifications
| Conjugate | Tb (Terbium) |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Microplate Reader |
| Quantity | 800 x 20 μL Assays |
| Packaging | 384-well Plate |
| Content And Storage | Store in ultra-cold freezer (-68 to -85°C). |
| Assay Entry | Biochemical coregulator interaction |
| Gene ID (Entrez) | 6257 |
| Readout | End Point |
| Shipping Condition | Dry Ice |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?