Learn More
Lapachol, TRC
Specifications
Specifications
| Chemical Name or Material | Lapachol |
| CAS | 84-79-7 |
| Molecular Formula | C15 H14 O3 |
| Quantity | 500 mg |
| Synonym | 1,4-Naphthalenedione, 2-hydroxy-3-(3-methyl-2-buten-1-yl)-, 1,4-Naphthalenedione, 2-hydroxy-3-(3-methyl-2-butenyl)- (9CI), 1,4-Naphthoquinone, 2-hydroxy-3-(3-methyl-2-butenyl)- (7CI,8CI), Lapachol (6CI), 2-Hydroxy-3-(3-methyl-2-buten-1-yl)-1,4-naphthalenedione, 2-Hydroxy-3-(3-methyl-2-butenyl)-1,4-naphthalenedione, 2-Hydroxy-3-(3-methyl-2-butenyl)-1,4-naphthoquinone, 2-Hydroxy-3-(3-methylbut-2-en-1-yl)-1,4-dihydronaphthalene-1,4-dione, Bethabarra Wood, C.I. 75490, C.I. Natural Yellow 16, Greenhartin, Ipe-tobacco Wood, Lapachol Wood, NSC 11905, NSC 629756, Surinam Greenheart Wood, Taigu Wood, Taiguic acid, Tecomin |
| InChI Formula | InChI=1S/C15H14O3/c1-9(2)7-8-12-13(16)10-5-3-4-6-11(10)14(17)15(12)18/h3-7,18H,8H2,1-2H3 |
| SMILES | CC(=CCC1=C(O)C(=O)c2ccccc2C1=O)C |
| IUPAC Name | 2-hydroxy-3-(3-methylbut-2-enyl)naphthalene-1,4-dione |
| Molecular Weight (g/mol) | 242.27 |
| Formula Weight | 242.0943 |