missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Larcaviximab Chimeric Recombinant Human Monoclonal Antibody

Larcaviximab Recombinant Human Monoclonal Antibody
Brand: Invitrogen MA542152
This item is not returnable.
View return policy
Description
Larcaviximab Recombinant Monoclonal Antibody for Western Blot, ELISA
Specifications
| Larcaviximab Chimeric | |
| Recombinant Monoclonal | |
| Unconjugated | |
| c4G7-N | |
| Ebola virus (Zaire). | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA, SDS-Page | |
| 1.53 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Human | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction