missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LARP1 Mouse anti-Human, Mouse, Rat, CoraLite 555, Clone: 2E9F2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL555-67810-100UL
This item is not returnable.
View return policy
Description
LARP1 Monoclonal antibody apecifically detetects LARP1 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.Specifications
| LARP1 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| LARP1 | |
| LARP1 fusion protein Ag29993 | |
| 100 μL | |
| 23367 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG1 |
| Immunofluorescence | |
| 2E9F2 | |
| CoraLite 555 | |
| BC033856 | |
| Mouse | |
| Protein G purification | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction