missing translation for 'onlineSavingsMsg'
Learn More

LAS-34823, TRC

Product Code. 30466056
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466056 100 mg 100mg
30464577 50 mg 50mg
30476300 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30466056 Supplier Toronto Research Chemicals Supplier No. TRCL395030100MG
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1708930-15-7
Molecular Formula C16H24BrNO2
Molecular Weight (g/mol) 342.27
Synonym LAS-34823, (R)-3-Hydroxy-1-(3-phenoxypropyl)-1-azoniabicyclo[2,2.2]octane Bromide, (R)-3-Hydroxy-1-(3-phenoxypropyl)quinuclidium Bromide
IUPAC Name (3R)-1-(3-phenoxypropyl)-1-azoniabicyclo[2.2.2]octan-3-ol;bromide
SMILES [Br-].O[C@H]1C[N+]2(CCCOc3ccccc3)CCC1CC2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H24NO2.BrH/c18-16-13-17(10-7-14(16)8-11-17)9-4-12-19-15-5-2-1-3-6-15;/h1-3,5-6,14,16,18H,4,7-13H2;1H/q+1;/p-1/t14?,16-,17?;/m0./s1
Formula Weight 341.099
Chemical Name or Material Aclidinium Bromide Impurity 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.