Learn More
Edmund Bühler GmbH™ Universal Tray for Platform Shakers
Product Code. 10665271
Change view
Click to view available options
For Use With (Equipment):
VKS 75
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10665271 VKS 75 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10665271 Supplier Edmund Bühler GmbH™ Supplier No. 0051 474
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Affix spring clamps or test tube racks through pre-drilled holes. Edmund Bühler GmbH™ Universal Tray for Platform Shakers require a hinged foot or test tube rack for affixing test tubes.
  • Fasten spring clamps, test tube racks, or holders for 1 microplate into pre-drilled holes
  • Maximum capacity for flasks: up to 2 x 5000mL
  • Maximum capacity for test tube racks: 5 pieces
  • For fastening test tube racks on a universal tray, test tube rack and hinged foot are necessary
Specifications
Holds Erlenmeyer Flasks, Test Tube Racks, Microplates
Product Type Universal Tray
Material Stainless Steel
Dimensions (L x W) 760 x 600 mm
Length (Metric) 760 mm
Width (Metric) 600 mm
For Use With (Equipment) VKS 75
Includes Drillings for Spring Clamps/Test Tube Racks