missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LBR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | LBR |
|---|---|
| Concentration | 0.21 mg/mL |
| Applications | Immunoprecipitation, Immunofluorescence, Flow Cytometry, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16810104
|
Proteintech
12398-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16800104
|
Proteintech
12398-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene belongs to the ERG4/ERG24 family. It localized in the nuclear envelope inner membrane and anchors the lamina and the heterochromatin to the membrane. It may mediate interaction between chromatin and lamin B. Mutations of this gene has been associated with autosomal recessive HEM/Greenberg skeletal dysplasia. Alternative splicing occurs at this locus and two transcript variants encoding the same protein have been identified.Specifications
| LBR | |
| Immunoprecipitation, Immunofluorescence, Flow Cytometry, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| O08984, Q14739, Q3U9G9 | |
| 3930, 89789, 98386 | |
| LBR Fusion Protein Ag3088 | |
| Primary | |
| -20°C | |
| LBR |
| 0.21 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| DHCR14B, lamin B receptor, LBR, LMN2R, PHA | |
| LBR | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title