Learn More
Trajan Scientific and Medical™ LC PEEKsil™ Tubing
Product Code. 15258763
Click to view available options
Overall Length (Metric):
50 mm
100 mm
150 mm
200 mm
500 mm
Diameter (Metric) Inner:
0.025 mm
0.05 mm
0.075 mm
0.1 mm
0.15 mm
0.175 mm
0.2 mm
0.3 mm
0.53 mm
Unit Size:
Pack of 2
Pack of 5
Product Code. 15258763
Brand: Trajan Scientific and Medical™ 0624293
Only null left
Add to basket
Add to basket
Mechanically robust polymer-sheathed fused silica tubing for use in HPLC systems.
  • Fused silica internal wall provides smooth surface, free of imperfections
  • PEEKsil compatible with most organic solvents and resistant to strong acids
  • Effective pH range of 0 to 10
  • Flexibility ensures easy connection between columns, detectors and injectors
    Specifications
    Type Tubing
    Includes 5/Pk.
    Diameter (Metric) Inner 0.075 mm
    Diameter (Metric) Outer 1.6 mm
    Overall Length (Metric) 200 mm
    Material Polymer-sheathed Fused Silica