missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3C Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18726-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to the propeptide of LC3C.
MAP1LC3C, also named as LC3C, belongs to the MAP1 LC3 family. It is the human homolog of yeast Atg8 and is involved in the formation of autophagosomal vacuoles, called autophagosomes. LC3C is produced by the proteolytic cleavage after the conserved C-terminal Gly residue. The antibody is specific to the prepeptide of LC3C.Specifications
| LC3C | |
| Polyclonal | |
| Unconjugated | |
| MAP1LC3C | |
| LC3C, MAP1A/MAP1B LC3 C, MAP1A/MAP1B light chain 3 C, MAP1LC3C | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 440738 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BXW4 | |
| MAP1LC3C | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction