missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LCHN Polyclonal Antibody
LCHN Polyclonal Antibody
Brand: Invitrogen PA597895
This item is not returnable.
View return policy
Description
LCHN Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
| LCHN | |
| Polyclonal | |
| Unconjugated | |
| DENND11 | |
| AI841796; DENN domain containing 11; DENN domain-containing protein 11; Dennd11; E330009J07Rik; KIAA1147; LCHN; mKIAA1147; predicted protein of HQ2561; PRO2561; protein LCHN; RGD1563986; RIKEN cDNA E330009J07 gene; si:ch211-238n5.4; similar to RIKEN cDNA E330009J07 gene; wu:fa55h08 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57189 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| A4D1U4 | |
| DENND11 | |
| Recombinant Human Protein LCHN protein (2-233AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction