Learn More
Thermo Scientific™ LDPE Foam-lined Polypropylene Caps for Bulk Separates
Product Code. 17830122
Change view
Click to view available options
Neck Finish:
24-410
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Finish Unit Size
17830122 24-410 Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17830122 Supplier Thermo Scientific™ Supplier No. 0672410WBP
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Use these LDPE foam-lined caps are for general-purpose HDPE bulk separate containers.
Thermo Scientific™ LDPE Foam-lined Polypropylene White Caps are for general-purpose HDPE bulk separate containers. The same high quality caps used on our Thermo Scientific certified HDPE environmental sample containers. Bottles sold separately (013-series and 012-series). Unprocessed and uncertified.
  • LDPE foam-lined white polypropylene cap
  • Bottles sold separately: 013-series and 012-series
  • Recommended for general purpose sampling where cleaning and certification is not required, or ready for your own cleaning and certification procedures
  • Economically bulk packed
Includes: Cap only
TRUSTED_SUSTAINABILITY
Specifications
Certification Parameters None
Color White
Description White PP Cap
Material Polypropylene
No. per Case 250
Septa N/A
Level Unprocessed
Liner Material LDPE Foam
Neck Finish 24-410
Shape Round
Certifications/Compliance Manufactured under an ISO certified quality management system.
Product Type Closure