Learn More
Fisherbrand™ Leaf Calibration Weight Set, Class F2 with certificate
Product Code. 15575109 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
1mg to 500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15575109 1mg to 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15575109
  • ONLINE EXCLUSIVE PRICE
    380.21€
    302.00€ / Pack of 1
    Save 78.21€
    20% Off
Estimated Shipment: 05-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use with weighing instruments of high accuracy class II. Fisherbrand™ Class F2 Stainless Steel Calibration Weights are also intended for use in important commercial transactions, such as those involving precious metals and stones.
  • Class F2 calibration weight
  • Weights intended for use in the verification or calibration of class M1 and possibly class M2 weights
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Classification F2
Weight (Metric) 1 to 500 mg
Mass 1 mg to 500 mg