Learn More
Tuttnauer Leakage Tray
Product Code. 17889588 Shop All Tuttnauer Products
Change view
Click to view available options
Dimensions (Dia. x H):
345 x 40 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (Dia. x H) unitSize
17889588 345 x 40 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17889588 Supplier Tuttnauer Supplier No. ZEA8300036
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Leakage Tray can be placed inside the wire basket to prevent the autoclave from getting dirty if there is a leakage of liquid from the bottles.
  • Leakage Tray to use under wire baskets to capture leaks.
  • Dimensions of the tray: diameter x H: 345 x 40 mm.
Specifications
Product Type Leak Tray
Dimensions (Dia. x H) 345 x 40 mm