Learn More
LENG8 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | LENG8 |
|---|---|
| Concentration | 507 μg/mL |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16821494
|
Proteintech
13336-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16811494
|
Proteintech
13336-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The LENG8 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human LENG8. This antibody recognizes human, mouse, rat antigen. The LENG8 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
| LENG8 | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG |
| 507 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 114823 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.