Learn More
Invitrogen™ Letaplimab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559228
 View more versions of this product
Product Code. 30283585
  • 1204.00€ / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Letaplimab is a monoclonal antibody that targets and inhibits the activity of the cytokine IL-22, thereby modulating immune responses.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Letaplimab
Recombinant Monoclonal
Unconjugated
IBI 188
Protein A
RUO
Human
Antibody
IgG4SP
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized