missing translation for 'onlineSavingsMsg'
Learn More

Leucomalachite Green-d6, TRC

Product Code. 30465346
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30465346 25 mg 25mg
30473664 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30465346 Supplier Toronto Research Chemicals Supplier No. TRCL33033325MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1173021-13-0
Molecular Formula C23 2H6 H20 N2
Molecular Weight (g/mol) 336.5
Synonym 4-[Deuterio-(4-dimethylaminophenyl)-(2,3,4,5,6-pentadeuteriophenyl)methyl]-N,N-dimethylaniline, Leucomalachite green D6, Leucomalachite green D6 (phenylmethin D6)
IUPAC Name 4-[deuterio-[4-(dimethylamino)phenyl]-(2,3,4,5,6-pentadeuteriophenyl)methyl]-N,N-dimethylaniline
SMILES [2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])(c2ccc(cc2)N(C)C)c3ccc(cc3)N(C)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C23H26N2/c1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4/h5-17,23H,1-4H3/i5D,6D,7D,8D,9D,23D
Formula Weight 336.2473
Percent Purity >95%
Chemical Name or Material Leucomalachite green D6
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.