missing translation for 'onlineSavingsMsg'
Learn More

Leucomycin Tartrate, TRC

Product Code. 30133375
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133375 500 mg 500mg
30133359 100 mg 100mg
17933675 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133375 Supplier Toronto Research Chemicals Supplier No. TRCL330328500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37280-56-1
Molecular Formula C36H63NO13.C4H6O6
Molecular Weight (g/mol) 867.97
Synonym Leucomycin (2R,3R)-2,3-dihydroxybutanedioate (salt) (9CI), Leucomycin [R-(R*,R*)]-2,3-dihydroxybutanedioate (salt), Kitasamycin tartrate, Trubin
IUPAC Name (9S,11E,13E,16S)-6-(((2S,4R,6R)-5-(((4R,6R)-4,5-dihydroxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-4-(dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-10-hydroxy-7-(2-hydroxyethyl)-4,5-dimethoxy-9,16-dimethyloxacyclohexadeca-11,13-dien-2-one (2R,3R)-2,3-dihydroxysuccinate
SMILES OC1[C@H](OC2C(CCO)C[C@H](C)C(O)/C=C/C=C/C[C@H](C)OC(CC(OC)C2OC)=O)O[C@H](C)C(OC3O[C@H](C)C(O)[C@@](O)(C)C3)[C@@H]1N(C)C.OC([C@H](O)[C@@H](O)C(O)=O)=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C36H63NO13.C4H6O6/c1-20-17-24(15-16-38)32(33(45-9)26(44-8)18-27(40)46-21(2)13-11-10-12-14-25(20)39)50-35-30(41)29(37(6)7)31(22(3)48-35)49-28-19-36(5,43)34(42)23(4)47-28;5-1(3(7)8)2(6)4(9)10/h10-12,14,20-26,28-35,38-39,41-43H,13,15-19H2,1-9H3;1-2,5-6H,(H,7,8)(H,9,10)/b11-10+,14-12+;/t20?,21?,22?,23?,24?,25?,26?,28?,29-,30?,31?,32?,33?,34?,35+,36?;1-,2-/m11/s1
Formula Weight 867.45
Chemical Name or Material Leucomycin Tartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.