missing translation for 'onlineSavingsMsg'
Learn More

Levetiracetam, TRC

Product Code. 17992855
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17992855 50 mg 50mg
17990755 100 mg 100mg
17964505 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17992855 Supplier Toronto Research Chemicals Supplier No. TRCL33150050MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102767-28-2
Molecular Formula C8 H14 N2 O2
Molecular Weight (g/mol) 170.21
Synonym 1-Pyrrolidineacetamide, α-ethyl-2-oxo-, (S)-, (αS)-α-Ethyl-2-oxo-1-pyrrolidineacetamide, (-)-(S)-α-Ethyl-2-oxo-1-pyrrolidineacetamide, (-)-Levetiracetam, (2S)-2-(2-Oxopyrrolidin-1-yl)butanamide, (S)-2-(2-Oxopyrrolidin-1-yl)butanamide, (S)-Levetiracetam, AGB 101, AGB 101 (low-dose levetiracetam), Keppra, Levesam, Levesam 500, Levetiracetam, Levipil, Levroxa, Matever, Spritam, UCB-L 059, Varuby
IUPAC Name (2S)-2-(2-oxopyrrolidin-1-yl)butanamide
SMILES CC[C@H](N1CCCC1=O)C(=O)N

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C8H14N2O2/c1-2-6(8(9)12)10-5-3-4-7(10)11/h6H,2-5H2,1H3,(H2,9,12)/t6-/m0/s1
Formula Weight 170.1055
Percent Purity >95%
Chemical Name or Material Levetiracetam
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.